Merck
  • [Simultaneous determination of cocaine and its metabolite ecgonine methyl ester in human blood using microwave extraction-gas chromatography].
[Simultaneous determination of cocaine and its metabolite ecgonine methyl ester in human blood using microwave extraction-gas chromatography].
Se pu = Chinese journal of chromatography (2010-11-05)
Xiaobo Wang, Nengsheng Ye, Jifen Wang, Xuexin Gu
ABSTRACT
A method was developed for the simultaneous determination of cocaine (COC) and its metabolite ecgonine methyl ester (EME) in human blood using gas chromatography-mass spectrometry (GC-MS) and gas chromatography-flame ionization detection (GC-FID). The blood sample was prepared by microwave extraction (MWE). The optimal parameters of MWE were as follows: 6 mL of chloroform-isopropanol (9: 1, v/v) mixture as extraction solvent, the pH value of the sample was adjusted at 10.0 with 0.05 mol/L Na2CO3-NaHCO3 buffer, the extraction was performed at 40 degrees C for 6 min. The COC and EME in the extract were qualified using GC-MS and quantitated using GC-FID. The average recoveries of COC and EME were from 79.91% to 99.85%, the relative standard deviations were less than 3.10%, and the limits of detection (LOD) were 60 and 40 mg/L, respectively. In the method COC and EME were detected without derivatization. The method is rapid, accurate and sensitive, and can be used for the simultaneous determination of COC and EME in blood samples.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Ecgonine methyl ester solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®