Merck
  • Orthostatic effects of midodrine versus L-NAME on cerebral blood flow and the renin-angiotensin-aldosterone system in tetraplegia.
Orthostatic effects of midodrine versus L-NAME on cerebral blood flow and the renin-angiotensin-aldosterone system in tetraplegia.
Archives of physical medicine and rehabilitation (2011-07-19)
Jill M Wecht, Miroslav Radulovic, Dwindally Rosado-Rivera, Run-Lin Zhang, Michael F LaFountaine, William A Bauman
ABSTRACT
To compare responses to head-up tilt (HUT) in individuals with chronic tetraplegia after midodrine hydrochloride (10 mg) versus nitro-L-arginine methyl ester (L-NAME, 1 mg/kg) administration. Prospective comparative drug trial. Veterans Affairs medical center. Participants (N=7) were studied during 3 laboratory visits: no drug, midodrine (administered orally 30 min before HUT), and L-NAME (infused over a 60-min period). Anti-hypotensive agents, midodrine, and L-NAME. Mean arterial pressure (MAP), cerebral blood flow (CBF), and markers of the renin-angiotensin-aldosterone system (RAAS, plasma renin and serum aldosterone) were measured in the supine position at baseline (BL) and during a 45° HUT maneuver. Data were compared between BL and the average of 3 assessments collected during HUT. Orthostatic MAP and CBF were increased with the midodrine and L-NAME groups compared with the no drug trial and the relationship between the change in MAP and CBF was significant (r=0.770; P<0.001). Both L-NAME and midodrine appeared to suppress the post-HUT RAAS response compared with no drug. Increasing orthostatic blood pressure with L-NAME or midodrine appears to increase CBF and suppress the RAAS during HUT in persons with tetraplegia, although more data are needed to confirm these preliminary findings.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Midodrine hydrochloride, analytical standard, for drug analysis