Merck
Comparison of ethyl glucuronide in hair with self-reported alcohol consumption.
Alcohol and alcoholism (Oxford, Oxfordshire) (2012-02-18)
Rosemary Lees, Robert Kingston, Tim M Williams, Graeme Henderson, Anne Lingford-Hughes, Matthew Hickman
ABSTRACT
Ethyl glucuronide (EtG) in hair is a proposed biomarker for alcohol consumption. This study compares hair EtG concentrations with self-reported alcohol consumption data, in individuals with a range of alcohol use. Hair was collected from 100 participants with a range of alcohol use. Participants completed an Alcohol Use Disorders Identification Test C questionnaire to record alcohol consumption. Participants were categorized into one of the four groups: tee-totallers (consuming 0 units a week), lower-risk drinkers (1-21 units a week), increasing-risk drinkers' consuming (22-50 units a week) and high-risk drinkers (over 50 units a week). Hair from the proximal 3 cm was analysed for EtG using gas chromatography-mass spectrometry/mass spectrometry. EtG was detected in 29 out of 100 hair samples. Based on the Society of Hair Testing (SOHT) threshold of 30 pg/mg EtG, the hair test identified alcohol consumption in 57.9% of high-risk drinkers, 45.5% of increasing-risk drinkers and only 9.8% of lower-risk drinkers. EtG sensitivity was highest for high-risk drinkers (consuming more than 50 units a week), identified to be 0.52 using a 30 pg/mg threshold and 0.58 using a 45 pg/mg threshold. A positive result is highly likely to indicate any drinking (positive predictive value, 1.00). A negative result does not provide good evidence for abstinence (negative predictive value, 0.23). EtG has been identified to be a low sensitivity marker that cannot be used quantitatively to determine alcohol exposure. EtG can be used qualitatively to indicate alcohol consumption with a positive result providing strong evidence for an individual drinking within the past 3 months.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Etil-β-D-glucuronide, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Etil-β-D-glucuronide, 100 μg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®