Merck
Semisynthesis of apigenin and acacetin-7-O-β-D-glycosides from naringin and their cytotoxic activities.
Carbohydrate research (2012-06-27)
Jidan Liu, Ling Chen, Shuanglian Cai, Qiuan Wang
ABSTRACT
Apigenin-7-O-β-D-glycosides 1-8 and acacetin-7-O-β-D-glycosides 9-16 were semisynthesized from 4'-O-benzyl apigenin 17 and acacetin 18 by glycosidation and deprotection with the corresponding α-acetylglycosyl bromide, respectively. Compounds 17 and 18 were prepared by iodination followed by base-induced elimination, 4'-O-benzylation, or 4'-O-methylation and acid hydrolysis using naringin as starting material which is readily available and cheap. Their cytotoxic potential against five human cancer cell lines (HL-60, SMMC-7721, A-549, MCF-7, and SW480) was evaluated by standard MTT method. The results show that compounds 2, 9, and 19 exhibit moderate cytotoxicity against HL-60, SMMC-7721, A-549, MCF-7, and SW480, while compound 3 exhibits potent cytotoxicity against MCF-7 selectively. Among the synthesized target compounds, 3, 4, 7, 11, 12, 15, and 16 were new compounds, the natural product 8 was the first synthesized and the synthesis of natural products 5, 6, 13, and 14 was efficiently improved by the new synthetic routes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acacetin, ≥97.0% (HPLC)