Merck
  • Pristimerin inhibits breast cancer cell migration by up- regulating regulator of G protein signaling 4 expression.
Pristimerin inhibits breast cancer cell migration by up- regulating regulator of G protein signaling 4 expression.
Asian Pacific journal of cancer prevention : APJCP (2012-07-18)
Xian-Min Mu, Wei Shi, Li-Xin Sun, Han Li, Yu-Rong Wang, Zhen-Zhou Jiang, Lu-Yong Zhang
ABSTRACT
Pristimerin isolated from Celastrus and Maytenus spp can inhibit proteasome activity. However, whether pristimerin can modulate cancer metastasis is unknown. The impacts of pristimerin on the purified and intracellular chymotrypsin proteasomal activity, the levels of regulator of G protein signaling 4 (RGS 4) expression and breast cancer cell lamellipodia formation, and the migration and invasion were determined by enzymatic, Western blot, immunofluorescent, and transwell assays, respectively. We found that pristimerin inhibited human chymotrypsin proteasomal activity in MDA-MB-231 cells in a dose-dependent manner. Pristimerin also inhibited breast cancer cell lamellipodia formation, migration, and invasion in vitro by up-regulating RGS4 expression. Thus, knockdown of RGS4 attenuated pristimerin-mediated inhibition of breast cancer cell migration and invasion. Furthermore, pristimerin inhibited growth and invasion of implanted breast tumors in mice. Pristmerin inhibits proteasomal activity and increases the levels of RGS4, inhibiting the migration and invasion of breast cancer cells.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Pristimerin
Prezzi e disponibilità al momento non sono disponibili