Merck
  • Dy(III)- and Yb(III)-curcuminoid compounds: original fluorescent single-ion magnet and magnetic near-IR luminescent species.
Dy(III)- and Yb(III)-curcuminoid compounds: original fluorescent single-ion magnet and magnetic near-IR luminescent species.
Chemistry (Weinheim an der Bergstrasse, Germany) (2012-08-02)
Melita Menelaou, Fatiha Ouharrou, Laura Rodríguez, Olivier Roubeau, Simon J Teat, Núria Aliaga-Alcalde
ABSTRACT
The multifunctional behavior of two mononuclear lanthanide compounds attached to a curcuminoid called 9 Accm has been investigated. The results show that [Dy(9 Accm)(2)(NO(3))(dmf)(2)] Yb(9 Accm)(3)(py)] behaves as a single-ion magnet and that both compounds display luminescent responses and exhibit affinity for graphite surfaces (see figure).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Dysprosium, ingot, 99.9% trace rare earth metals basis
Sigma-Aldrich
Dysprosium, chips, 99.9% trace rare earth metals basis