Merck
  • Pymetrozine is hydroxylated by CYP6CM1, a cytochrome P450 conferring neonicotinoid resistance in Bemisia tabaci.
Pymetrozine is hydroxylated by CYP6CM1, a cytochrome P450 conferring neonicotinoid resistance in Bemisia tabaci.
Pest management science (2013-01-18)
Ralf Nauen, John Vontas, Martin Kaussmann, Katharina Wölfel
ABSTRACT
Resistance to neonicotinoid insecticides such as imidacloprid in the cotton whitefly, Bemisia tabaci, is linked to its hydroxylation by constitutively overexpressed CYP6CM1, a cytochrome P450 enzyme. Here, an investigation was conducted to establish whether CYP6CM1 functionally expressed in Sf9 cells also detoxifies pymetrozine, a selective homopteran feeding blocker known to be cross-resistant to neonicotinoids in whiteflies. Incubation of pymetrozine with functionally expressed Bemisia CYP6CM1 and subsequent LC-MS/MS analysis revealed a rapid formation of two pymetrozine metabolites by hydroxylation of its heterocyclic 1,2,4-triazine ring system. Enzyme kinetics revealed a Km value of 5.9 ± 0.3 µM and a time-dependent depletion of pymetrozine. The known cross-resistance between imidacloprid, other neonicotinoid insecticides and pymetrozine in B. tabaci is most likely conferred by the very same detoxification mechanism, i.e. a monooxygenase-based hydroxylation mechanism linked to the overexpression of CYP6CM1. These insecticide chemistries should not be alternated in whitefly resistance management strategies.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Pymetrozin, PESTANAL®, analytical standard