Merck
Pediatric intensive care treatment of uncontrolled status epilepticus.
Critical care clinics (2013-03-30)
Ryan Wilkes, Robert C Tasker
ABSTRACT
The critically ill mechanically ventilated child with ongoing seizures that are refractory to any treatment presents a distinct challenge in pediatric neurocritical care. The evidence base from randomized controlled trials on which anti-epileptic drug (AED) strategy should be used is inadequate. This review of refractory and super-refractory status epilepticus summarizes recent pediatric case series regarding definitions, the second-tier AED therapies once initial anticonvulsants have failed, and the experience of high-dose midazolam, barbiturate anesthesia, and volatile anesthetics for uncontrolled status epilepticus.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Midazolam solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®