Merck
Metabolism and pharmacokinetic studies of propionylpromazine in horses.
Journal of chromatography (1989-04-14)
J Park, Y O Shin, H Y Choo
ABSTRACT
The propionylpromazine concentrations in plasma after intramuscular administration to horses were determined using gas chromatography with nitrogen-phosphorus detection. After hydrolysis by beta-glucuronidase/arylsulphatase, the parent drug and three metabolites were detected in urine. The metabolites were identified as 2-(1-hydroxypropyl)promazine, 2-(1-propenyl)promazine and 7-hydroxypropionylpromazine by gas chromatography-mass spectrometry. No N-demethylated or sulphoxidated metabolites of propionylpromazine were observed in the horse urine.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Propionylpromazine hydrochloride