Merck
A resonance Raman study on the structures of complexes of flavoprotein D-amino acid oxidase.
Journal of biochemistry (1986-02-01)
Y Nishina, R Miura, H Tojo, Y Miyake, H Watari, K Shiga
ABSTRACT
Resonance Raman (RR) spectra were obtained for the purple complexes of D-amino acid oxidase (DAO) with D-lysine or N-methylalanine. RR spectra of a complex of oxidized DAO with the oxidation product of D-lysine or D-proline were also measured. The isotope shifts of the observed bands of the purple complex with D-lysine upon 13C- or 15N-substitution of lysine indicate that the ligand is delta 1-piperideine-2-carboxylate. That the band at 1671 cm-1 for the purple intermediate with N-methylalanine shifts to 1666 cm-1 in D2O solution indicates that the imino acid, N-methyl-alpha-iminopropionate, has a protonated imino group. Many bands due to a ligand in the RR spectra of the complex of oxidized DAO with an oxidation product can be observed below 1000 cm-1, but no band for the purple complex is seen in this frequency region. The band associated with the CO2-symmetric stretching mode of the product, such as delta 1-piperideine-2-carboxylate or delta 1-pyrrolidine-2-carboxylate, complexed with the oxidized DAO shifts in D2O solution. This suggests that the product imino acid interacts with the enzyme through some proton(s).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-Methyl-L-alanine, ≥98.0% (TLC)