Merck
  • The 5-lipoxygenase inhibitor BWA4C impairs osteoclastic resorption in a synchronized model of bone remodeling.
The 5-lipoxygenase inhibitor BWA4C impairs osteoclastic resorption in a synchronized model of bone remodeling.
Bone (1995-08-01)
C Franchi-Miller, J L Saffar
ABSTRACT
The role of leukotrienes on bone resorption was tested in a well-standardized model of bone remodeling by inhibiting their biosynthesis with BWA4C, a specific inhibitor of 5-lipoxygenase. After extraction of their upper molars unilaterally, 30 Wistar rats were divided into three groups; the first remained untreated (control group), the second received 80 mg/kg/day of BWA4C dissolved in polyethylene glycol 300 (experimental group), and the third received only the vehicle (sham-treated group). After four days of experiment, the animals were killed and the resorption profile was assessed along the antagonist mandibular buccal cortex. The main result was a dramatic decrease in the number of TRAP-positive mononucleated preosteoclasts in the experimental group (-69%, p < 0.0005 and p < 0.003 vs. the control and sham-treated groups, respectively). This drop was related to a significant decrease in the number of osteoclasts. Neither the activation of the differentiated osteoclasts nor their mean interface with the bone surface were affected by BWA4C. Concomitantly, the mast cell population residing near the vascular network limiting the periosteum was markedly and significantly increased by the treatment. These mast cells were mostly degranulating, i.e., were in a state of activation that we previously found to related to resorption. These data suggest (1) that the leukotrienes are involved in the recruitment of osteoclast progenitors, and/or their differentiation into preosteoclasts, and (2) that mast cells responded to leukotriene inhibition.(ABSTRACT TRUNCATED AT 250 WORDS)
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
BW A4C, ≥98% (HPLC)