- Synthesis and activity of 1-N-iminosugar inhibitors, siastatin B analogues for alpha-N-acetylgalactosaminidase and beta-N-acetylglucosaminidase.
Synthesis and activity of 1-N-iminosugar inhibitors, siastatin B analogues for alpha-N-acetylgalactosaminidase and beta-N-acetylglucosaminidase.
Bioorganic & medicinal chemistry (1996-01-01)
Y Nishimura, T Satoh, T Kudo, S Kondo, T Takeuchi
PMID8689244
ABSTRACT
N-Acetylgalactosamine-based 1-N-iminosugars, new types of glycosidase inhibitor have been synthesized by modeling on siastatin B, isolated from a Streptomyces culture. The analogues of siastatin B were proved to be potent inhibitors for alpha-N-acetylgalactosaminidase and/or beta-N-acetylglucosaminidase.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto