Merck
A case of lethal intoxication after ingestion of toquilone compositum.
The American journal of forensic medicine and pathology (1996-09-01)
N Fucci
ABSTRACT
A case of acute intoxication of both methaqualone and diphenhydramine is reported. The analysis of these compounds was performed by liquid-liquid extraction (Toxi-Lab DPC procedure) followed by gas chromatography/mass spectrometry determination; both substances are contained in the pharmaceutical formulation called Toquilone Compositum (Medichemie, Switzerland).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Methaqualone solution, 1 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®