- Synthesis and CNS activity of new 1-alkyl-2-aryl-7-hydroxy-5(1H)oxo-imidazo[1,2-a]pyrymidines.
Synthesis and CNS activity of new 1-alkyl-2-aryl-7-hydroxy-5(1H)oxo-imidazo[1,2-a]pyrymidines.
Acta poloniae pharmaceutica (1996-05-01)
D Matosiuk, T Tkaczyński, J Stefańczyk
PMID8960291
ABSTRACT
Cyclocondensation of 1,5-disubstituted 2-aminoimidazolines with diethyl malonate in the presence of sodium methanolate leading to imidazol[1,2-alpyrimidine-5,7-dion system is presented. Prevailing of 7-hydroxy-5(1H)oxo form between three possible tautomers is discussed based on spectral data. CNS activity of nine compounds is evaluated.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Prezzi e disponibilità al momento non sono disponibili