Merck
Distribution of cyclic imide-transforming activity in microorganisms.
FEMS microbiology letters (1998-02-07)
C L Soong, J Ogawa, H Sukiman, T Prana, M Sri Prana, S Shimizu
ABSTRACT
Cyclic imide-transforming activity was found to be widely distributed in bacteria, yeast and molds. This activity was not correlated with cyclic ureide-transforming activity in bacteria, but there was some correlation in yeast and molds. These two activities are probably catalyzed by different enzymes in bacteria. Besides the well-known cyclic ureide transformation, cyclic imide transformation by microorganisms was common.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
5-Methylhydantoin, 97%