Merck
Parathyroid hormone-induced lipolysis in human adipose tissue.
Journal of lipid research (1987-05-01)
A Taniguchi, K Kataoka, T Kono, F Oseko, H Okuda, I Nagata, H Imura
ABSTRACT
Relative lipolytic activity of human parathyroid hormone-(1-34) (hPTH-(1-34], hPTH-(3-34), desamino-Ser1-hPTH-(1-34), and rat PTH-(1-34) was compared in human subcutaneous adipose tissues in vitro. Human PTH-(1-34), rat PTH-(1-34), and desamino-Ser1-hPTH-(1-34) stimulated in vitro lipolysis significantly above basal level at the concentration of 10(-6) M. Average increments of lipolytic rate were 2.39, 1.82, and 0.87 mumol/g per 2 hr, respectively, being significantly different among the three groups. On the other hand, hPTH-(3-34)-induced lipolytic rate was 0.83 +/- 0.18 mumol/g per 2 hr, not significantly different from the basal level (0.71 +/- 0.20 mumol/g per 2 hr). The effect of hPTH-(3-34) on glycerol release stimulated by hPTH-(1-34), isoproterenol, or forskolin was subsequently investigated. Human PTH-(3-34) produced a dose-dependent inhibition of hPTH-(1-34)-stimulated lipolysis. In contrast, isoproterenol- and forskolin-induced lipolytic rates were not influenced by hPTH-(3-34). The effect of propranolol on hPTH-(1-34)- or isoproterenol-induced lipolysis was also studied. Propranolol dose-dependently inhibited isoproterenol-induced lipolysis but had no effect on lipolysis stimulated by hPTH-(1-34). These results suggest that the amino acids at positions 1 (serine) and 2 (valine) of PTH are critical for the stimulation of lipolysis in human adipose tissue. Human PTH-(1-34) causes lipolysis after binding to receptors distinct from beta-adrenergic receptors of fat cells and possibly hPTH-(3-34) inhibits hPTH-(1-34)-stimulated lipolysis by competing at the level of PTH receptor.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
D-Serine, ≥98% (TLC)