Merck
Sept4/ARTS is required for stem cell apoptosis and tumor suppression.
Genes & development (2010-10-19)
María García-Fernández, Holger Kissel, Samara Brown, Travis Gorenc, Andrew J Schile, Shahin Rafii, Sarit Larisch, Hermann Steller
ABSTRACT
Inhibitor of Apoptosis Proteins (IAPs) are frequently overexpressed in tumors and have become promising targets for developing anti-cancer drugs. IAPs can be inhibited by natural antagonists, but a physiological requirement of mammalian IAP antagonists remains to be established. Here we show that deletion of the mouse Sept4 gene, which encodes the IAP antagonist ARTS, promotes tumor development. Sept4-null mice have increased numbers of hematopoietic stem and progenitor cells, elevated XIAP protein, increased resistance to cell death, and accelerated tumor development in an Eμ-Myc background. These phenotypes are partially suppressed by inactivation of XIAP. Our results suggest that apoptosis plays an important role as a frontline defense against cancer by restricting the number of normal stem cells.