Merck
  • Expression of the vanin gene family in normal and inflamed human skin: induction by proinflammatory cytokines.
Expression of the vanin gene family in normal and inflamed human skin: induction by proinflammatory cytokines.
The Journal of investigative dermatology (2009-03-27)
Patrick A M Jansen, Marijke Kamsteeg, Diana Rodijk-Olthuis, Ivonne M J J van Vlijmen-Willems, Gys J de Jongh, Mieke Bergers, Geuranne S Tjabringa, Patrick L J M Zeeuwen, Joost Schalkwijk
ABSTRACT
The vanin gene family encodes secreted and membrane-bound ectoenzymes that convert pantetheine into pantothenic acid and cysteamine. Recent studies in a mouse colitis model indicated that vanin-1 has proinflammatory activity and suggest that pantetheinases are potential therapeutic targets in inflammatory diseases. In a microarray analysis of epidermal gene expression of psoriasis and atopic dermatitis lesions, we identified vanin-3 as the gene showing the highest differential expression of all annotated genes that we studied (19-fold upregulation in psoriasis). Quantitative real-time PCR analysis confirmed the microarray data on vanin-3 and showed similar induction of vanin-1, but not of vanin-2, in psoriatic epidermis. Immunohistochemistry showed that vanin-3 is expressed in the differentiated epidermal layers. Using submerged and organotypic keratinocyte cultures, we found that vanin-1 and vanin-3 are induced at the mRNA and protein level by psoriasis-associated proinflammatory cytokines (Th17/Th1) but not by Th2 cytokines. We hypothesize that increased levels of pantetheinase activity are part of the inflammatory-regenerative epidermal differentiation program, and may contribute to the phenotype observed in psoriasis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Anti-VNN3 antibody produced in rabbit, affinity isolated antibody