Merck
  • Constituents of essential oils from the leaves, stem barks and resins of Canarium parvum Leen., and Canarium tramdenanum Dai et Yakovl. (Burseracea) grown in Vietnam.
Constituents of essential oils from the leaves, stem barks and resins of Canarium parvum Leen., and Canarium tramdenanum Dai et Yakovl. (Burseracea) grown in Vietnam.
Natural product research (2014-01-22)
Tran D Thang, Do N Dai, Ngo X Luong, Isiaka A Ogunwande
ABSTRACT
The chemical constituents of essential oils from the leaf, stem bark and resins of Canarium parvum Leen., and Canarium tramdenanum Dai et Yakovl. (Burseracea) grown in Vietnam are being reported. The hydrodistilled oils were analysed for their chemical constituents by means of gas chromatography-flame ionisation detector and gas chromatography coupled with mass spectrometry. The main compounds of C. parvum were β-caryophyllene (18.7%), (E)-β-ocimene (12.9%), (Z)-β-ocimene (11.9%), germacrene D (8.8%) and α-humulene (8.4%) in the leaf; β-caryophyllene (30.4%), α-copaene (20.5%) and (E)-β-ocimene (7.7%) in the stem. However, germacrene D (23.2%), α-amorphene (14.9%), α-copaene (9.8%) and β-elemene (8.6%) were present in the resin. The leaf of C. tramdenanum comprises β-caryophyllene (16.8%), α-phellandrene (15.9%), γ-elemene (13.1%) and limonene (11.8%), while limonene (25.7%), α-phellandrene (21.7%), α-pinene (12.3%) and β-caryophyllene (10.9%) were present in the stem. However, δ-elemene (14.6%) and bulnesol (16.0%) are the main constituents in the resin.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
α-Pinene, 98%