Merck
Circadian clock NAD+ cycle drives mitochondrial oxidative metabolism in mice.
Science (New York, N.Y.) (2013-09-21)
Clara Bien Peek, Alison H Affinati, Kathryn Moynihan Ramsey, Hsin-Yu Kuo, Wei Yu, Laura A Sena, Olga Ilkayeva, Biliana Marcheva, Yumiko Kobayashi, Chiaki Omura, Daniel C Levine, David J Bacsik, David Gius, Christopher B Newgard, Eric Goetzman, Navdeep S Chandel, John M Denu, Milan Mrksich, Joseph Bass
ABSTRACT
Circadian clocks are self-sustained cellular oscillators that synchronize oxidative and reductive cycles in anticipation of the solar cycle. We found that the clock transcription feedback loop produces cycles of nicotinamide adenine dinucleotide (NAD(+)) biosynthesis, adenosine triphosphate production, and mitochondrial respiration through modulation of mitochondrial protein acetylation to synchronize oxidative metabolic pathways with the 24-hour fasting and feeding cycle. Circadian control of the activity of the NAD(+)-dependent deacetylase sirtuin 3 (SIRT3) generated rhythms in the acetylation and activity of oxidative enzymes and respiration in isolated mitochondria, and NAD(+) supplementation restored protein deacetylation and enhanced oxygen consumption in circadian mutant mice. Thus, circadian control of NAD(+) bioavailability modulates mitochondrial oxidative function and organismal metabolism across the daily cycles of fasting and feeding.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥95% (HPLC)
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥99%
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, suitable for cell culture, ≥96.5% (HPLC), ≥96.5% (spectrophotometric assay), from yeast
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, Grade AA-1
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 10 mg (per vial)
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, ≥98%, BioUltra, from yeast
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 50 mg (per vial)
Sigma-Aldrich
β-Nicotinamide adenine dinucleotide, pkg of 20 mg (per vial)
Sigma-Aldrich
β-Dinucleotide nicotinamide adenina, purified by column chromatography, ≥99%