Merck
  • Plasminogen activator inhibitor-1 antagonist TM5441 attenuates Nω-nitro-L-arginine methyl ester-induced hypertension and vascular senescence.
Plasminogen activator inhibitor-1 antagonist TM5441 attenuates Nω-nitro-L-arginine methyl ester-induced hypertension and vascular senescence.
Circulation (2013-10-05)
Amanda E Boe, Mesut Eren, Sheila B Murphy, Christine E Kamide, Atsuhiko Ichimura, David Terry, Danielle McAnally, Layton H Smith, Toshio Miyata, Douglas E Vaughan
ABSTRACT
Long-term inhibition of nitric oxide synthase by L-arginine analogues such as N(ω)-nitro-l-arginine methyl ester (L-NAME) has been shown to induce senescence in vitro and systemic hypertension and arteriosclerosis in vivo. We previously reported that plasminogen activator inhibitor-1 (PAI-1)-deficient mice (PAI-1(-/-)) are protected against L-NAME-induced pathologies. In this study, we investigated whether a novel, orally active PAI-1 antagonist (TM5441) has a similar protective effect against L-NAME treatment. Additionally, we studied whether L-NAME can induce vascular senescence in vivo and investigated the role of PAI-1 in this process. Wild-type mice received either L-NAME or L-NAME and TM5441 for 8 weeks. Systolic blood pressure was measured every 2 weeks. We found that TM5441 attenuated the development of hypertension and cardiac hypertrophy compared with animals that had received L-NAME alone. Additionally, TM5441-treated mice had a 34% reduction in periaortic fibrosis relative to animals on L-NAME alone. Finally, we investigated the development of vascular senescence by measuring p16(Ink4a) expression and telomere length in aortic tissue. We found that L-NAME increased p16(Ink4a) expression levels and decreased telomere length, both of which were prevented with TM5441 cotreatment. Pharmacological inhibition of PAI-1 is protective against the development of hypertension, cardiac hypertrophy, and periaortic fibrosis in mice treated with L-NAME. Furthermore, PAI-1 inhibition attenuates the arterial expression of p16(Ink4a) and maintains telomere length. PAI-1 appears to play a pivotal role in vascular senescence, and these findings suggest that PAI-1 antagonists may provide a novel approach in preventing vascular aging and hypertension.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Nω-Nitro-L-arginine methyl ester hydrochloride, ≥97% (TLC), powder