Merck
  • Calmodulin antagonists inhibit insulin-stimulated GLUT4 (glucose transporter 4) translocation by preventing the formation of phosphatidylinositol 3,4,5-trisphosphate in 3T3L1 adipocytes.
Calmodulin antagonists inhibit insulin-stimulated GLUT4 (glucose transporter 4) translocation by preventing the formation of phosphatidylinositol 3,4,5-trisphosphate in 3T3L1 adipocytes.
Molecular endocrinology (Baltimore, Md.) (2000-02-16)
C Yang, R T Watson, J S Elmendorf, D B Sacks, J E Pessin
ABSTRACT
It has been previously reported that calmodulin plays a regulatory role in the insulin stimulation of glucose transport. To examine the basis for this observation, we examined the effect of a panel of calmodulin antagonists that demonstrated a specific inhibition of insulin-stimulated glucose transporter 4 (GLUT4) but not insulin- or platelet-derived growth factor (PDGF)-stimulated GLUT1 translocation in 3T3L1 adipocytes. These treatments had no effect on insulin receptor autophosphorylation or tyrosine phosphorylation of insulin receptor substrate 1 (IRS1). Furthermore, IRS1 or phosphotyrosine antibody immunoprecipitation of phosphatidylinositol (PI) 3-kinase activity was not affected. Despite the marked insulin and PDGF stimulation of PI 3-kinase activity, there was a near complete inhibition of protein kinase B activation. Using a fusion protein of the Grp1 pleckstrin homology (PH) domain with the enhanced green fluorescent protein, we found that the calmodulin antagonists prevented the insulin stimulation of phosphatidylinositol 3,4,5-trisphosphate [PI(3,4,5)P3] formation in vivo. Similarly, although PDGF stimulation increased PI 3-kinase activity in in vitro immunoprecipitation assays, there was also no significant formation of PI(3,4,5)P3 in vivo. These data demonstrate that calmodulin antagonists prevent insulin-stimulated GLUT4 translocation by inhibiting the in vivo production of PI(3,4,5)P3 without directly affecting IRS1- or phosphotyrosine-associated PI 3-kinase activity. This phenomenon is similar to that observed for the PDGF stimulation of 3T3L1 adipocytes.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
N-(6-Aminohexyl)-1-naphthalenesulfonamide hydrochloride, ≥98% (HPLC)