Merck
[The anti-ischemic effect of O, O-acetyldaurisoline].
Yao xue xue bao = Acta pharmaceutica Sinica (2002-04-10)
T Wang, H Mu, G Liu
ABSTRACT
O, O-acetyldaurisoline(Adau) is a derivative of daurisoline (Dau). In cultured PC12 cells, Adau was found to inhibit K+, Bay K8644 and norepinephrine induced intracellular free calcium concentration increase. Adau was also shown to protect PC12 cells from hypoxia-reoxygenation injury with EC50 of 64.3 +/- 12.5 mumol.L-1 by MTT assay. Adau (2.5, 5 and 10 mg.kg-1, i.v.) administrated 30 min before ischemia in bilateral carotid artery occlusion and four-vessel occlusion rats, attenuated the increase of lipid peroxide content and the decrease of SOD activity. These results show that Adau have significant protective effects against ischemic injury.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Daurisoline, ≥98% (HPLC)