Merck
  • Effects of gentiopicroside, sweroside and swertiamarine, secoiridoids from gentian (Gentiana lutea ssp. symphyandra), on cultured chicken embryonic fibroblasts.
Effects of gentiopicroside, sweroside and swertiamarine, secoiridoids from gentian (Gentiana lutea ssp. symphyandra), on cultured chicken embryonic fibroblasts.
Planta medica (2006-03-25)
Nilgün Oztürk, Seval Korkmaz, Yusuf Oztürk, K Hüsnü Can Başer
ABSTRACT
Wound healing properties of Gentian (Gentiana lutea ssp. symphyandra) extract and its main constituents, gentiopicroside, sweroside and swertiamarine (compounds 1-3, respectively) were evaluated by comparison with dexpanthenol on cultured chicken embryonic fibroblasts. The extract was also analyzed by HPLC to quantify its constituents. Chicken embryonic fibroblasts from fertilized eggs were incubated with the plant extract and its constituents, compounds 1-3. Using microscopy, mitotic ability, morphological changes and collagen production in the cultured fibroblasts were evaluated as parameters. Wound healing activity of Gentian seems to be mainly due to the increase in the stimulation of collagen production and the mitotic activity by compounds 2 and 3, respectively (p < 0.005 in all cases). All three compounds also exhibited cytoprotective effects, which may cause a synergism in terms of wound healing activity of Gentian. The findings demonstrated the wound healing activity of Gentian, which has previously been based only on ethnomedical data.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sweroside, ≥95% (LC/MS-ELSD)