Merck
  • Enhanced dissolution and bioavailability of raloxifene hydrochloride by co-grinding with different superdisintegrants.
Enhanced dissolution and bioavailability of raloxifene hydrochloride by co-grinding with different superdisintegrants.
Chemical & pharmaceutical bulletin (2010-03-02)
Balasubramaniam Jagadish, Rajesh Yelchuri, Bindu K, Hemalatha Tangi, Swetha Maroju, Vinay Umesh Rao
ABSTRACT
The present study investigated the effect of co-grinding raloxifene HCL (RHCL) with different superdisintegrants, namely crospovidone (CP), croscarmellose sodium (CCS) and sodium starch glycolate (SSG), using a ball mill, in order to determine the potential effect on dissolution rate and bioavailability of raloxifene hydrochloride (RHCL). The dissolution studies of the co-ground compositions and the corresponding physical mixtures were carried out in U.S. Pharmacopeia (USP) Type II apparatus. The solid state interactions of the co-ground and the physical mixtures were evaluated by differential scanning calorimetry (DSC) and X-ray diffraction (XRD). The pharmacokinetics of co-ground mixture (1 : 5 RHCL : CP) and milled RHCL was evaluated following oral administration (25 mg/kg) in healthy female Sprague-Dawley rats. DSC studies showed that the crystalline nature of RHCL was reduced after co-grinding with superdisintegrants, while co-grinding with CP resulted in significant particle-size reduction of the mixture. Significant enhancement in dissolution rate was observed with co-ground mixture of RHCL with CP (1 : 5). The extent of the mean plasma exposures of RHCL was 7-fold higher in animals treated with co-ground mixture of RHCL, CP (1 : 5) compared to animals treated with milled RHCL. Co-grinding of RHCL with CP, reduced drug crystallinity, increased the rate and extent of dissolution, and improved bioavailability.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
USP
Sodium Starch Glycolate Type A, United States Pharmacopeia (USP) Reference Standard