Merck
  • Different manufacturers of syringes: a new source of variability in blood gas, acid-base balance and related laboratory test?
Different manufacturers of syringes: a new source of variability in blood gas, acid-base balance and related laboratory test?
Clinical biochemistry (2012-03-24)
Gabriel Lima-Oliveira, Giuseppe Lippi, Gian Luca Salvagno, Martina Montagnana, Geraldo Picheth, Gian Cesare Guidi
ABSTRACT
Evaluate whether four different brands/types of heparin coated syringes can represent a source of variability in blood gas analysis (BGA). Blood was collected from one hundred volunteers into different syringes: Syringe I (lithium heparin and calcium balanced); Syringe II: in-house prepared (sodium heparin); Syringe III: (spray-dried calcium-balanced lithium heparin); Syringe IV (lyophilized electrolyte-balanced lithium heparin). Significant differences were as follows: a) Syringe I vs II: pO(2), sO(2), pCO(2)(t), cHCO(3)(-), ctCO(2), base excess (BE), total hemoglobin (tHb), sodium (Na(+)), potassium (K(+)), calcium (Ca(2+)), glucose (Glu), lactate (Lac), O(2) Hb and p 50; b) Syringe I vs III: pH, pO(2), cHCO(3)(-), ctCO(2), BE, Na(+), Glu, Lac and p 50; c) Syringe I vs IV: pH, pO(2), sO(2), pCO(2)(t), BE, Na(+), K(+), Ca(2+), Glu, Lac and O(2) Hb; d) Syringe II vs III: pH, pO(2), sO(2), pCO(2)(t), cHCO(3)(-), ctCO(2), ctO(2), tHb, Na(+), K(+), Ca(2+), Lac and p 50; e) Syringe II vs IV: pH, pO(2), sO(2), pCO(2)(t), cHCO(3)(-), ctCO(2), BE, tHb, Na(+), K(+), Ca(2+), Lac, O(2) Hb and p 50; f) Syringe III vs IV: pH, pO(2), sO(2), cHCO(3)(-), ctCO(2), ctO(2), BE, Na(+), K(+), Ca(2+), O(2) Hb and p 50. The different manufacturers of syringes can represent new source of variability on BGA.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Heparin calcium for NMR identification, European Pharmacopoeia (EP) Reference Standard