Merck
  • Novel (coumarin-3-yl)carbamates as selective MAO-B inhibitors: synthesis, in vitro and in vivo assays, theoretical evaluation of ADME properties and docking study.
Novel (coumarin-3-yl)carbamates as selective MAO-B inhibitors: synthesis, in vitro and in vivo assays, theoretical evaluation of ADME properties and docking study.
European journal of medicinal chemistry (2013-03-12)
Maria J Matos, Santiago Vilar, Rosa Ma Gonzalez-Franco, Eugenio Uriarte, Lourdes Santana, Carol Friedman, Nicholas P Tatonetti, Dolores Viña, Jose A Fontenla
ABSTRACT
A series of (coumarin-3-yl)carbamates was synthesized and evaluated in vitro as monoamine oxidase (MAO-A and MAO-B) inhibitors. Most of the new compounds selectively inhibited MAO-B isoenzyme with IC50 values in the micro or nanoMolar ranges. Since these compounds must achieve the brain cells, theoretical evaluation of ADME properties were also carried out. Compound 8 (benzyl(coumarin-3-yl)carbamate), which presented the most interesting in vitro MAO-B inhibitory profile (IC50 against MAO-B = 45 nM), was subjected to further studies. This in vitro MAO-B inhibitory activity is comparable with that of the selegiline, the reference compound (IC50 against MAO-B = 20 nM). Taking into account the in vitro results of compound 8, in vivo assays and docking calculations were also carried out for this derivative.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
(RS)-Selegiline hydrochloride, European Pharmacopoeia (EP) Reference Standard