Merck
Calcitonin gene-related peptide receptors in human gastrointestinal epithelia.
British journal of pharmacology (1994-12-01)
H M Cox, I R Tough
ABSTRACT
1. The secretory responses to calcitonin gene-related peptide (CGRP) receptor agonists have been characterized in two human adenocarcinoma cell lines, namely HCA-7 and Colony-29 (Col-29) epithelia. These cells form polarized epithelial layers when grown on permeable supports and allow changes in electrogenic ion transport in response to agonists to be monitored continuously. 2. alpha-CGRP (rat and human sequences), rat beta-CGRP and human [Tyr0]CGRP applied to the basolateral surface were found to be full agonists, causing prolonged increases in short-circuit current. Concentration-response curves exhibited EC50 values of 0.6-1.5 nM in HCA-7 cells. The same agonists were less effective in Col-29 epithelia, the EC50 values ranging from 1 to 10 nM in these cells. [Cys(ACM)2,7]CGRP was effective in both cell lines and was more potent in HCA-7 cells. 3. CGRP receptors were preferentially located on the basolateral surface in both cell types. Addition of r alpha-CGRP to the apical domain produced significantly smaller secretory responses (8.1% in HCA-7 and 29.2% in Col-29) compared with those produced following basolateral application (100%). 4. In both cell lines r alpha-CGRP-elevated short-circuit current was inhibited by the loop diuretic piretanide (200 microM) and by somatostatin (100 nM). Pretreating epithelia with the cyclo-oxygenase inhibitor, piroxicam (5 microM) had no significant effect upon CGRP responses in either cell line. 5. Rat alpha-CGRP (0.2 nM) responses in HCA-7 epithelia were inhibited by the C-terminal fragment CGRP(8-37) (1 microM). Pretreatment of Col-29 cells with CGRP(8-37) did not, however, alter the size or profile of responses to r alpha-CGRP (1 nM).6. We conclude that high-affinity CGRP receptors exist on the basolateral surface of both cell lines,however they differ in their sensitivity to CGRP(8-37) and agonist orders of potency. Thus different CGRP receptor subtypes appear to predominate in these two epithelial cell types.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Piretanide, European Pharmacopoeia (EP) Reference Standard
Piretanide for system suitability, European Pharmacopoeia (EP) Reference Standard