Merck
[Immune stimulative potency of milk proteins].
Polski merkuriusz lekarski : organ Polskiego Towarzystwa Lekarskiego (2014-04-12)
Adam Ambroziak, Grazyna Cichosz
ABSTRACT
Milk proteins are characterized by the highest immune stimulative potency from among all the proteins present in human diet. Whey proteins and numerous growth factors that regulate insulin secretion, differentiation of intestine epithelium cells, and also tissue restoration, are priceless in stimulation the immune system. Lactoferrin shows the most comprehensive pro-health properties: antioxidative, anticancer, immune stimulative and even chemopreventive. Also peptides and amino acids formed from casein and whey proteins possess immune stimulative activity. The most valuable proteins, i.e. lactoferrin, immune globulins, lactoperoxidase and lisozyme, together with bioactive peptides, are resistant to pepsin and trypsin activity. This is why they maintain their exceptional biological activity within human organism. Properly high consumption of milk proteins conditions correct function of immune system, especially at children and elderly persons.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Lisozima, powder or granules, ≥39,000 units/mg protein
Sigma-Aldrich
Lisozima, BioUltra, lyophilized powder, ≥98% (SDS-PAGE), ≥40,000 units/mg protein
Sigma-Aldrich
Lysozyme human, Lysobac, recombinant, expressed in rice, lyophilized powder, ≥100,000 units/mg protein
Sigma-Aldrich
Lisozima, powder (crystalline), 70000-140000 U/mg
Sigma-Aldrich
Lisozima, 10 mg/mL
Sigma-Aldrich
Lysozyme chloride form from chicken egg white, Grade VI, ≥35,000 units/mg protein (E1%/282)
Sigma-Aldrich
Lisozima, dialyzed, lyophilized, powder, ~100000 U/mg
Sigma-Aldrich
Lisozima, For use as a marker in SDS-PAGE
Sigma-Aldrich
Lisozima, aseptically filled
Sigma-Aldrich
Lysozyme from human neutrophils, ≥95% (SDS-PAGE), lyophilized powder, ≥100,000 units/mg protein (E1%/280)
Supelco
Lisozima, VETRANAL®, analytical standard