Merck
Structural basis for inactivation of Giardia lamblia carbamate kinase by disulfiram.
The Journal of biological chemistry (2014-02-22)
Andrey Galkin, Liudmila Kulakova, Kap Lim, Catherine Z Chen, Wei Zheng, Illarion V Turko, Osnat Herzberg
ABSTRACT
Carbamate kinase from Giardia lamblia is an essential enzyme for the survival of the organism. The enzyme catalyzes the final step in the arginine dihydrolase pathway converting ADP and carbamoyl phosphate to ATP and carbamate. We previously reported that disulfiram, a drug used to treat chronic alcoholism, inhibits G. lamblia CK and kills G. lamblia trophozoites in vitro at submicromolar IC50 values. Here, we examine the structural basis for G. lamblia CK inhibition of disulfiram and its analog, thiram, their activities against both metronidazole-susceptible and metronidazole-resistant G. lamblia isolates, and their efficacy in a mouse model of giardiasis. The crystal structure of G. lamblia CK soaked with disulfiram revealed that the compound thiocarbamoylated Cys-242, a residue located at the edge of the active site. The modified Cys-242 prevents a conformational transition of a loop adjacent to the ADP/ATP binding site, which is required for the stacking of Tyr-245 side chain against the adenine moiety, an interaction seen in the structure of G. lamblia CK in complex with AMP-PNP. Mass spectrometry coupled with trypsin digestion confirmed the selective covalent thiocarbamoylation of Cys-242 in solution. The Giardia viability studies in the metronidazole-resistant strain and the G. lamblia CK irreversible inactivation mechanism show that the thiuram compounds can circumvent the resistance mechanism that renders metronidazole ineffectiveness in drug resistance cases of giardiasis. Together, the studies suggest that G. lamblia CK is an attractive drug target for development of novel antigiardial therapies and that disulfiram, an FDA-approved drug, is a promising candidate for drug repurposing.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-cisteina, from non-animal source, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
L-cisteina, 97%
Sigma-Aldrich
L-cisteina, BioUltra, ≥98.5% (RT)
Sigma-Aldrich
Metronidazole, BioXtra
Supelco
Metronidazole, analytical standard
SAFC
L-cisteina
Supelco
Metronidazole, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-cisteina, ≥97%, FG
Sigma-Aldrich
Tetraethylthiuram disulfide, ≥97.0% (S)
Supelco
L-cisteina, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-cisteina, Wacker Chemie AG, ≥98.0%
Disulfiram, European Pharmacopoeia (EP) Reference Standard
Supelco
Metronidazole, VETRANAL®, analytical standard
Metronidazole, European Pharmacopoeia (EP) Reference Standard