Merck
  • Molecular mechanistic explanation for the spectrum of cholestatic disease caused by the S320F variant of ABCB4.
Molecular mechanistic explanation for the spectrum of cholestatic disease caused by the S320F variant of ABCB4.
Hepatology (Baltimore, Md.) (2014-05-09)
Edward J Andress, Michael Nicolaou, Marta R Romero, Sandhia Naik, Peter H Dixon, Catherine Williamson, Kenneth J Linton
ABSTRACT
ABCB4 flops phosphatidylcholine into the bile canaliculus to protect the biliary tree from the detergent activity of bile salts. Homozygous-null ABCB4 mutations cause the childhood liver disease, progressive familial intrahepatic cholestasis, but cause and effect is less clear, with many missense mutations linked to less severe cholestatic diseases. ABCB4(S320F), in particular, is described in 13 patients, including in heterozygosity with ABCB4(A286V), ABCB4(A953D), and null mutants, whose symptoms cover the spectrum of cholestatic disease. We sought to define the impact of these mutations on the floppase, explain the link with multiple conditions at the molecular level, and investigate the potential for reversal. ABCB4(S320F), ABCB4(A286V), and ABCB4(A953D) expression was engineered in naïve cultured cells. Floppase expression, localization, and activity were measured by western blot, confocal microscopy, and lipid transport assays, respectively. ABCB4(S320F) was fully active for floppase activity but expression at the plasma membrane was reduced to 50%. ABCB4(A286V) expressed and trafficked efficiently but could not flop lipid, and ABCB4(A953D) expressed poorly and was impaired in floppase activity. Proteasome inhibition stabilized nascent ABCB4(S320F) and ABCB4(A953D) but did not improve plasma membrane localization. Cyclosporin-A improved plasma membrane localization of both ABCB4(S320F) and ABCB4(A953D), but inhibited floppase activity. The level of ABCB4 functionality correlates with, and is the primary determinant of, cholestatic disease severity in these patients. ABCB4(S320F) homozygosity, with half the normal level of ABCB4, is the tipping point between more benign and potentially fatal cholestasis and makes these patients more acutely sensitive to environmental effects. Cyclosporin-A increased expression of ABCB4(S320F) and ABCB4(A953D), suggesting that chemical chaperones could be exploited for therapeutic benefit to usher in a new era of personalized medicine for patients with ABCB4-dependent cholestatic disease.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodio cloruro, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodio cloruro, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodio cloruro, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodio cloruro, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
SAFC
Sodio cloruro, 5 M
Sigma-Aldrich
Soluzione tampone HEPES, 1 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
Sodio cloruro, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Cyclosporin A, 97.0-101.5% (on dried basis)
Supelco
Sodio cloruro, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, ≥95% (HPLC), solid
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Supelco
Cyclosporine, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodio cloruro, 0.85%
Sigma-Aldrich
Sodio cloruro, tested according to Ph. Eur.
Sigma-Aldrich
Cyclosporin A, from Tolypocladium inflatum, BioReagent, Molecular Biology, ≥95%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%