Merck
Sucralfate and Helicobacter pylori.
Journal of gastroenterology (1996-11-01)
A Terano, T Shimada, H Hiraishi
ABSTRACT
Sucralfate enhances the eradication rate of Helicobacter pylori when administered with antibiotics. Sucralfate acts on H. pylori through mucus gel. Whether sucralfate exerts its anti-ulcer action through the eradication of H. pylori is still controversial. As sucralfate itself is an effective, well-tolerated and cost-effective anti-ulcer agent, its mechanisms of action should be investigated further.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Sucrose octasulfate–aluminum complex
Sucralfate, European Pharmacopoeia (EP) Reference Standard