Merck
Amprenavir.
Drugs (1998-06-09)
J C Adkins, D Faulds
ABSTRACT
Amprenavir is a viral protease inhibitor with specificity for the HIV protease enzyme. The resistance profile of amprenavir appears to differ from that of other protease inhibitors such as saquinavir and indinavir. Twelve hours after single-dose administration of amprenavir 1200mg to HIV-infected individuals, the mean plasma concentration of the drug was more than 10-fold greater than the 50% inhibitory concentration for HIV-1IIIB in peripheral blood lymphocytes. In a small nonblind study, amprenavir monotherapy increased CD4+ cell count and decreased viral load in 37 patients with HIV infection and no previous exposure to protease inhibitor therapy. Combination therapy comprising amprenavir and other antiretroviral agents (abacavir, zidovudine, lamivudine, indinavir, saquinavir or nelfinavir) decreased viral load and increased CD4+ cell counts in patients with HIV infection. Antiviral efficacy was maintained during up to 24 weeks' follow-up. Available data suggest that rash, headache and diarrhoea or loose stools are the most frequent adverse events associated with amprenavir therapy.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Amprenavir, ≥98% (HPLC)
Prezzi e disponibilità al momento non sono disponibili