Merck
GH administration rescues fatty liver regeneration impairment by restoring GH/EGFR pathway deficiency.
Endocrinology (2014-04-09)
A Collin de l'Hortet, A Zerrad-Saadi, C Prip-Buus, V Fauveau, N Helmy, M Ziol, C Vons, K Billot, V Baud, Hélène Gilgenkrantz, Jacques-Emmanuel Guidotti
ABSTRACT
GH pathway has been shown to play a major role in liver regeneration through the control of epidermal growth factor receptor (EGFR) activation. This pathway is down-regulated in nonalcoholic fatty liver disease. Because regeneration is known to be impaired in fatty livers, we wondered whether a deregulation of the GH/EGFR pathway could explain this deficiency. Hepatic EGFR expression and triglyceride levels were quantified in liver biopsies of 32 obese patients with different degrees of steatosis. We showed a significant inverse correlation between liver EGFR expression and the level of hepatic steatosis. GH/EGFR down-regulation was also demonstrated in 2 steatosis mouse models, a genetic (ob/ob) and a methionine and choline-deficient diet mouse model, in correlation with liver regeneration defect. ob/ob mice exhibited a more severe liver regeneration defect after partial hepatectomy (PH) than methionine and choline-deficient diet-fed mice, a difference that could be explained by a decrease in signal transducer and activator of transcription 3 phosphorylation 32 hours after PH. Having checked that GH deficiency accounted for the GH signaling pathway down-regulation in the liver of ob/ob mice, we showed that GH administration in these mice led to a partial rescue in hepatocyte proliferation after PH associated with a concomitant restoration of liver EGFR expression and signal transducer and activator of trnascription 3 activation. In conclusion, we propose that the GH/EGFR pathway down-regulation is a general mechanism responsible for liver regeneration deficiency associated with steatosis, which could be partially rescued by GH administration.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Choline chloride, ≥98%
Sigma-Aldrich
L-metionina, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Sigma-Aldrich
Choline chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥98%
Sigma-Aldrich
Choline chloride, ≥99%
SAFC
L-metionina
Sigma-Aldrich
L-metionina, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-metionina, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Choline chloride, BioUltra, ≥99.0% (AT)
Supelco
L-metionina, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Choline chloride, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Choline chloride, United States Pharmacopeia (USP) Reference Standard
L-metionina, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Somatotropin from human pituitary, recombinant, expressed in E. coli, ≥95% (SDS-PAGE), lyophilized powder
Supelco
L-metionina, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
Human GH ELISA Kit, for serum, plasma, cell culture supernatant and urine