Merck
  • Cutaneous exposure scenarios for engineered nanoparticles used in semiconductor fabrication: a preliminary investigation of workplace surface contamination.
Cutaneous exposure scenarios for engineered nanoparticles used in semiconductor fabrication: a preliminary investigation of workplace surface contamination.
International journal of occupational and environmental health (2014-07-08)
Michele Shepard, Sara Brenner
ABSTRACT
Numerous studies are ongoing in the fields of nanotoxicology and exposure science; however, gaps remain in identifying and evaluating potential exposures from skin contact with engineered nanoparticles in occupational settings. The aim of this study was to identify potential cutaneous exposure scenarios at a workplace using engineered nanoparticles (alumina, ceria, amorphous silica) and evaluate the presence of these materials on workplace surfaces. Process review, workplace observations, and preliminary surface sampling were conducted using microvacuum and wipe sample collection methods and transmission electron microscopy with elemental analysis. Exposure scenarios were identified with potential for incidental contact. Nanoparticles of silica or silica and/or alumina agglomerates (or aggregates) were identified in surface samples from work areas where engineered nanoparticles were used or handled. Additional data are needed to evaluate occupational exposures from skin contact with engineered nanoparticles; precautionary measures should be used to minimize potential cutaneous exposures in the workplace.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
LUDOX® AS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
LUDOX® HS-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Silice, nanopowder, 10-20 nm particle size (BET), 99.5% trace metals basis
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <25 nm particle size (BET)
Sigma-Aldrich
LUDOX® TM-50 colloidal silica, 50 wt. % suspension in H2O
Sigma-Aldrich
Cerium(IV) oxide, powder, <5 μm, 99.9% trace metals basis
Sigma-Aldrich
Silice, nanoparticles, mesoporous, 200 nm particle size, pore size 4 nm
Sigma-Aldrich
Silice, nanopowder (spherical, porous), 5-20 nm particle size (TEM), 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
LUDOX® HS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silice, nanopowder, 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Cerium(IV) oxide, ≥99.0%
Sigma-Aldrich
LUDOX® SM colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Silice, MCM-41 type (hexagonal)
Sigma-Aldrich
LUDOX® TM-40 colloidal silica, 40 wt. % suspension in H2O
Sigma-Aldrich
Cerium(IV) oxide, nanopowder, <50 nm particle size (BET), 99.95% trace rare earth metals basis
Sigma-Aldrich
Silice
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
LUDOX® AS-30 colloidal silica, 30 wt. % suspension in H2O
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh