Merck
Management of patients with active caries.
Journal of the California Dental Association (2014-08-01)
Peter Milgrom
ABSTRACT
This paper reports on a mechanism to manage caries as a disease and to medically intervene in the disease process to halt progression. The goal of this paper is to provide this alternative to a surgical-only approach. The management of caries begins with assessing lesion activity and the potential for arrest. This requires a clinical and radiological assessment and evaluation of risk. Hopeless teeth are extracted and large cavities filled to reduce infection. Risk reduction strategies are employed so efforts to arrest lesions can be successful. Teeth with lesions in the enamel or outer third of the dentin should be sealed, not restored, as restorations can weaken teeth and can be traumatic to pulps.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Poly(vinylpyrrolidone)–Iodine complex
Povidone (iodinated), European Pharmacopoeia (EP) Reference Standard