Merck
An unexpected journey: lysine methylation across the proteome.
Biochimica et biophysica acta (2014-02-25)
Kaitlyn E Moore, Or Gozani
ABSTRACT
The dynamic modification of histone proteins by lysine methylation has emerged over the last decade as a key regulator of chromatin functions. In contrast, our understanding of the biological roles for lysine methylation of non-histone proteins has progressed more slowly. Though recently it has attracted less attention, ε-methyl-lysine in non-histone proteins was first observed over 50 years ago. In that time, it has become clear that, like the case for histones, non-histone methylation represents a key and common signaling process within the cell. Recent work suggests that non-histone methylation occurs on hundreds of proteins found in both the nucleus and the cytoplasm, and with important biomedical implications. Technological advances that allow us to identify lysine methylation on a proteomic scale are opening new avenues in the non-histone methylation field, which is poised for dramatic growth. Here, we review historical and recent findings in non-histone lysine methylation signaling, highlight new methods that are expanding opportunities in the field, and discuss outstanding questions and future challenges about the role of this fundamental post-translational modification (PTM).
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
L-lisina, ≥98% (TLC)
Sigma-Aldrich
L-lisina, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-lisina, crystallized, ≥98.0% (NT)
Sigma-Aldrich
L-lisina, reagent grade, ≥98% (HPLC)
Supelco
L-lisina, analytical standard
Supelco
L-lisina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-lisina, BioUltra, ≥99.5% (AT)
L-lisina, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Supelco
L-lisina, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material
Lysine acetate, European Pharmacopoeia (EP) Reference Standard