Merck
Reductive dehalogenase structure suggests a mechanism for B12-dependent dehalogenation.
Nature (2014-10-21)
Karl Ap Payne, Carolina P Quezada, Karl Fisher, Mark S Dunstan, Fraser A Collins, Hanno Sjuts, Colin Levy, Sam Hay, Stephen Ej Rigby, David Leys
ABSTRACT
Organohalide chemistry underpins many industrial and agricultural processes, and a large proportion of environmental pollutants are organohalides. Nevertheless, organohalide chemistry is not exclusively of anthropogenic origin, with natural abiotic and biological processes contributing to the global halide cycle. Reductive dehalogenases are responsible for biological dehalogenation in organohalide respiring bacteria, with substrates including polychlorinated biphenyls or dioxins. Reductive dehalogenases form a distinct subfamily of cobalamin (B12)-dependent enzymes that are usually membrane associated and oxygen sensitive, hindering detailed studies. Here we report the characterization of a soluble, oxygen-tolerant reductive dehalogenase and, by combining structure determination with EPR (electron paramagnetic resonance) spectroscopy and simulation, show that a direct interaction between the cobalamin cobalt and the substrate halogen underpins catalysis. In contrast to the carbon-cobalt bond chemistry catalysed by the other cobalamin-dependent subfamilies, we propose that reductive dehalogenases achieve reduction of the organohalide substrate via halogen-cobalt bond formation. This presents a new model in both organohalide and cobalamin (bio)chemistry that will guide future exploitation of these enzymes in bioremediation or biocatalysis.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Vitamina B12, ≥98%
Supelco
Vitamina B12, pharmaceutical secondary standard, certified reference material
Sigma-Aldrich
Vitamina B12, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥98%
Supelco
Vitamina B12, analytical standard
Sigma-Aldrich
Cobalt, powder, 2 μm particle size, 99.8% trace metals basis
Supelco
Vitamina B12, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Cobalt, powder, <150 μm, ≥99.9% trace metals basis
Sigma-Aldrich
Vitamina B12, meets USP testing specifications
Sigma-Aldrich
Cobalt, granular, 99.99% trace metals basis
Sigma-Aldrich
Cobalt, Carbon coated magnetic, nanopowder, <50 nm particle size (TEM), ≥99%
Sigma-Aldrich
Cobalt, pieces, 99.5% trace metals basis
Sigma-Aldrich
2,6-Dibromophenol, 99%
Vitamina B12, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Cobalt, wire, diam. 1.0 mm, 99.995% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 1.0 mm, 99.95% trace metals basis
Sigma-Aldrich
Cobalt, foil, thickness 0.1 mm, ≥99.99%
Sigma-Aldrich
Cobalt, rod, diam. 5.0 mm, 99.95% trace metals basis
Cobalt, foil, light tested, 25x25mm, thickness 0.01mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.05mm, annealed, 99.9%
Cobalt, foil, 8mm disks, thickness 0.003mm, 99.9%
Cobalt, foil, 6mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 4mm disks, thickness 0.007mm, 99.9%
Cobalt, foil, 8mm disks, thickness 0.01mm, 99.9%
Cobalt, foil, 25x25mm, thickness 0.25mm, as rolled, 99.9%
Cobalt, foil, light tested, 25x25mm, thickness 0.05mm, as rolled, 99.99+%
Cobalt, foil, light tested, 50x50mm, thickness 0.0125mm, 99.9%
Cobalt, wire reel, 0.5m, diameter 0.05mm, hard, 99.9%
Cobalt, foil, 25mm disks, thickness 0.125mm, annealed, 99.9%
Cobalt, foil, 8mm disks, thickness 0.006mm, 99.9%