Merck
  • Chronic alcohol consumption leads to neurochemical changes in the nucleus accumbens that are not fully reversed by withdrawal.
Chronic alcohol consumption leads to neurochemical changes in the nucleus accumbens that are not fully reversed by withdrawal.
Neurotoxicology and teratology (2014-06-04)
Pedro A Pereira, João Neves, Manuel Vilela, Sérgio Sousa, Catarina Cruz, M Dulce Madeira
ABSTRACT
Neuropeptide Y (NPY)- and acetylcholine-containing interneurons of the nucleus accumbens (NAc) seem to play a major role in the rewarding effects of alcohol. This study investigated the relationship between chronic alcohol consumption and subsequent withdrawal and the expression of NPY and acetylcholine in the NAc, and the possible involvement of nerve growth factor (NGF) in mediating the effects of ethanol. Rats ingesting an aqueous ethanol solution over 6months and rats subsequently deprived from ethanol during 2months were used to estimate the total number and the somatic volume of NPY and cholinergic interneurons, and the numerical density of cholinergic varicosities in the NAc. The tissue content of choline acetyltransferase (ChAT) and catecholamines were also determined. The number of NPY interneurons increased during alcohol ingestion and returned to control values after withdrawal. Conversely, the number and the size of cholinergic interneurons, and the amount of ChAT were unchanged in ethanol-treated and withdrawn rats, but the density of cholinergic varicosities was reduced by 50% during alcohol consumption and by 64% after withdrawal. The concentrations of dopamine and norepinephrine were unchanged both during alcohol consumption and after withdrawal. The administration of NGF to withdrawn rats significantly increased the number of NPY-immunoreactive neurons, the size of cholinergic neurons and the density of cholinergic varicosities. Present data show that chronic alcohol consumption leads to long-lasting neuroadaptive changes of the cholinergic innervation of the NAc and suggest that the cholinergic system is a potential target for the development of therapeutic strategies in alcoholism and abstinence.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido perclorico, ACS reagent, 70%
Sigma-Aldrich
Acido perclorico, 70%, 99.999% trace metals basis
Sigma-Aldrich
Acido perclorico, ACS reagent, 60%
Sigma-Aldrich
Methylene blue, certified by the BSC
Sigma-Aldrich
Methylene Blue solution, concentrate according to Ehrlich, concentrated, aqueous solution
Sigma-Aldrich
Acido perclorico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., 70.0-72.0%
Sigma-Aldrich
Methylene Blue solution, 0.05 wt. % in H2O
Sigma-Aldrich
Methylene Blue solution, suitable for microscopy
Sigma-Aldrich
Methylene Blue solution, suitable for microbiology
Sigma-Aldrich
Methylene Blue Solution 1.4%(w/v)95%ethanol | 7220-79-3, 1.4 % (w/v) in 95% ethanol
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
Sigma-Aldrich
Acido perclorico, puriss. p.a., ACS reagent, packed in coated, shock- and leak-protected glass bottle, ≥60% (T)