Merck
  • Three-Component O-Demethylase System Essential for Catabolism of a Lignin-Derived Biphenyl Compound in Sphingobium sp. Strain SYK-6.
Three-Component O-Demethylase System Essential for Catabolism of a Lignin-Derived Biphenyl Compound in Sphingobium sp. Strain SYK-6.
Applied and environmental microbiology (2014-09-14)
Taichi Yoshikata, Kazuya Suzuki, Naofumi Kamimura, Masahiro Namiki, Shojiro Hishiyama, Takuma Araki, Daisuke Kasai, Yuichiro Otsuka, Masaya Nakamura, Masao Fukuda, Yoshihiro Katayama, Eiji Masai
ABSTRACT
Sphingobium sp. strain SYK-6 is able to assimilate lignin-derived biaryls, including a biphenyl compound, 5,5'-dehydrodivanillate (DDVA). Previously, ligXa (SLG_07770), which is similar to the gene encoding oxygenase components of Rieske-type nonheme iron aromatic-ring-hydroxylating oxygenases, was identified to be essential for the conversion of DDVA; however, the genes encoding electron transfer components remained unknown. Disruption of putative electron transfer component genes scattered through the SYK-6 genome indicated that SLG_08500 and SLG_21200, which showed approximately 60% amino acid sequence identities with ferredoxin and ferredoxin reductase of dicamba O-demethylase, were essential for the normal growth of SYK-6 on DDVA. LigXa and the gene products of SLG_08500 (LigXc) and SLG_21200 (LigXd) were purified and were estimated to be a trimer, a monomer, and a monomer, respectively. LigXd contains FAD as the prosthetic group and showed much higher reductase activity toward 2,6-dichlorophenolindophenol with NADH than with NADPH. A mixture of purified LigXa, LigXc, and LigXd converted DDVA into 2,2',3-trihydroxy-3'-methoxy-5,5'-dicarboxybiphenyl in the presence of NADH, indicating that DDVA O-demethylase is a three-component monooxygenase. This enzyme requires Fe(II) for its activity and is highly specific for DDVA, with a Km value of 63.5 μM and kcat of 6.1 s(-1). Genome searches in six other sphingomonads revealed genes similar to ligXc and ligXd (>58% amino acid sequence identities) with a limited number of electron transfer component genes, yet a number of diverse oxygenase component genes were found. This fact implies that these few electron transfer components are able to interact with numerous oxygenase components and the conserved LigXc and LigXd orthologs are important in sphingomonads.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Vanillina, ReagentPlus®, 99%
Supelco
Vanillina, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Vanillina, natural, ≥97%, FCC, FG
Sigma-Aldrich
Vanillina, ≥97%, FCC, FG
USP
Vanillina, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Vanillina, tested according to Ph. Eur.
Supelco
Vanillina, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Vanillina, traceable to primary standards (LGC)
Supelco
Digoxigenin, analytical standard
Supelco
Vanillina, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Vanillina, European Pharmacopoeia (EP) Reference Standard
Digoxigenin, European Pharmacopoeia (EP) Reference Standard