Merck
  • Quantification of intracellular and extracellular digoxin and ouabain by liquid chromatography/electrospray ionization tandem mass spectrometry.
Quantification of intracellular and extracellular digoxin and ouabain by liquid chromatography/electrospray ionization tandem mass spectrometry.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2014-12-03)
Hiroaki Yamaguchi, Kazuaki Miyamori, Toshihiro Sato, Jiro Ogura, Masaki Kobayashi, Takehiro Yamada, Nariyasu Mano, Ken Iseki
ABSTRACT
A liquid chromatography/tandem mass spectrometry method for the determination of intracellular accumulation in addition to transcellular transport of digoxin and ouabain in renal epithelial HK-2 cells was developed. The solid-phase extraction Bond Elut(®) C18 (100mg/1mL) cartridge was used for the extraction of digoxin and ouabain from extracellular (medium) and intracellular (cell lysate) matrices. Chromatographic separation was performed on a CAPCELL PAK C18 MGII column (2.0mm×150mm, 5μm). This method covered a linear range of 0.5-1000ng/mL of concentrations in medium and 0.5-1000ng of concentrations in cell lysate for digoxin and ouabain. The intra-day precision and inter-day precision of analysis were less than 11.9%, and the accuracy was within ±11.6%. The total run time was 16min. Our method was successfully applied to the transport experiments of digoxin and ouabain by HK-2 cell monolayers.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Ammonio acetato, ACS reagent, ≥97%
Sigma-Aldrich
Desametasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Ammonio acetato, ≥99.99% trace metals basis
Supelco
Ammonio acetato, LiChropur, eluent additive for LC-MS
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Ammonio acetato, Molecular Biology, ≥98%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Desametasone, ≥98% (HPLC), powder
Sigma-Aldrich
Metanolo, Absolute - Acetone free
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
USP
Metanolo, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Metanolo, BioReagent, ≥99.93%
Supelco
Metanolo, analytical standard
Sigma-Aldrich
Ammonio acetato, 99.999% trace metals basis
Supelco
Metanolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Ammonio acetato, reagent grade, ≥98%
Sigma-Aldrich
Ammonio acetato, BioXtra, ≥98%
Supelco
Desametasone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Desametasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
Ammonium acetate solution, BioUltra, ~5 M in H2O
USP
Desametasone, United States Pharmacopeia (USP) Reference Standard