Merck
Prognostic significance of MSI2 predicts unfavorable outcome in adult B-acute lymphoblastic leukemia.
International journal of laboratory hematology (2014-08-06)
R M Aly, H F Ghazy
ABSTRACT
The Musashi-2 gene (MSI2) is implicated in leukemogenesis, and high MSI2 expression has been associated with decreased survival in acute myeloid leukemia (AML) and acute lymphoid leukemia (ALL), suggesting its use as a new prognostic marker. We aimed to validate the prognostic significance of MSI2 in ALL. MSI2 expression was measured by real-time polymerase chain reaction in 140 adult B-ALL patients and compared to controls. MSI2 expression level in patients was significantly higher when compared to the control group (P = 0.001). High MSI2 expression did not correlate with the clinical characteristics of patients. However, patients with high MSI2 expression had significantly lower incidence of complete remission (CR) (P = 0.03), inferior overall survival (P = 0.018), and shorter disease-free survival (P = 0.001). Multivariate analysis revealed that high MSI2 expression was an independent prognostic factor for adult BCR-ABL1-negative B-ALL patients. These results confirm the association of MSI2 expression with outcome in adult B-ALL and demonstrate the utility of MSI2 as a clinical prognostic biomarker.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acqua, suitable for HPLC
Sigma-Aldrich
Acqua, Nuclease-Free Water, for Molecular Biology
Sigma-Aldrich
Acqua, sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acqua, HPLC Plus
Sigma-Aldrich
Acqua, Deionized
Supelco
Acqua, suitable for ion chromatography
Sigma-Aldrich
Acqua, for embryo transfer, sterile-filtered, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Acqua, Molecular Biology, sterile filtered
Supelco
Acqua, for TOC analysis
Sigma-Aldrich
Acqua, BioPerformance Certified
Acqua, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Sigma-Aldrich
Acqua, ACS reagent
Supelco
Acqua, for ultratrace analysis, ACS reagent
Sigma-Aldrich
Acqua, for cell biology, sterile ultrafiltered
Sigma-Aldrich
Acqua, PCR Reagent
Sigma-Aldrich
Acqua, tested according to Ph. Eur.
Supelco
Acqua, suitable for HPCE, suitable for luminescence, suitable for UV/Vis spectroscopy
Sigma-Aldrich
Acqua, endotoxin, free
Acqua, UKAS ISO/IEC17025 and ISO Guide 34 certified, density: 0.9982 g/mL at 20 °C, density: 0.9970 g/mL at 25 °C
Supelco
Acqua, H&D Fitzgerald Ltd. Quality
Sigma-Aldrich
Water-16O, ≥99.94 atom % 16O