Merck
  • Impact of prolonged-release oxycodone/naloxone on outcomes affecting patients' daily functioning in comparison with extended-release tapentadol: a systematic review.
Impact of prolonged-release oxycodone/naloxone on outcomes affecting patients' daily functioning in comparison with extended-release tapentadol: a systematic review.
Clinical therapeutics (2015-01-17)
Deepika Thakur, Sara Dickerson, Mohit Kumar Bhutani, Rod Junor
ABSTRACT
The objective of this systematic review was to assess the clinical efficacy, safety, tolerability, and health-related quality of life outcomes associated with management of moderate-to-severe chronic pain with oxycodone/naloxone and tapentadol, focusing on the effect of these treatments on patients' daily functioning. Literature from a wide range of sources, including Embase, MEDLINE, MEDLINE In-Process, and the Cochrane Central Register of Controlled Trials, was searched to identify randomized controlled trials investigating tapentadol or oxycodone/naloxone for the treatment of patients with chronic pain. A network meta-analysis was conducted to determine the relative efficacy and safety profiles of these treatments. Oxycodone/naloxone was significantly better than tapentadol with respect to the Patient Assessment of Constipation Symptoms total score (risk ratio = -3.60; 95% credible interval, -5.36 to -2.11) and revealed a significantly lower risk of dizziness (risk ratio = 0.72; 95% credible interval, 0.42-0.98). Oxycodone/naloxone was directionally favored, although not significantly superior to tapentadol for headache, fatigue, dry mouth, dyspepsia, and withdrawals due to lack of efficacy. For the AE outcomes of constipation, nausea, and vomiting, as well as pain efficacy and all-cause withdrawals from studies, tapentadol was directionally favored without any statistical difference from oxycodone/naloxone. However, the two treatments were not wholly comparable for the evaluation of pain efficacy because of differences in on-study rescue medication and a higher baseline pain severity in the tapentadol studies. Oxycodone/naloxone offers significant improvements in Patient Assessment of Constipation Symptoms total score and dizziness and was directionally favored for fatigue and headache compared with extended-release tapentadol, which may translate to improved patient daily functioning and health-related quality of life.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Supelco
Acetaminofene, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetylsalicylic acid, ≥99.0%
Supelco
Aspirin (Acetyl Salicylic Acid), Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetaminofene, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetaminofene, BioXtra, ≥99.0%
Sigma-Aldrich
Aspirin, meets USP testing specifications
Sigma-Aldrich
Acetaminofene, meets USP testing specifications, 98.0-102.0%, powder
Sigma-Aldrich
Acetaminofene, analytical standard
USP
Aspirin, United States Pharmacopeia (USP) Reference Standard
Acetaminofene, European Pharmacopoeia (EP) Reference Standard
USP
Naloxone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetylsalicylic acid, analytical standard
Supelco
Acetaminophen solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Acetylsalicylic acid, European Pharmacopoeia (EP) Reference Standard
Acetylsalicylic acid for peak identification, European Pharmacopoeia (EP) Reference Standard