Merck
  • Caffeic acid inhibits the uptake of 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) by inducing the efflux transporters expression in Caco-2 cells.
Caffeic acid inhibits the uptake of 2-amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) by inducing the efflux transporters expression in Caco-2 cells.
Biological & pharmaceutical bulletin (2014-11-18)
Yun-Jin Hong, Sung-Yong Yang, Mi-Hyun Nam, Yun-Chang Koo, Kwang-Won Lee
ABSTRACT
2-Amino-1-methyl-6-phenylimidazo[4,5-b]pyridine (PhIP) is formed as a by-product of the Maillard reaction during cooking and frying of protein-rich foods at high temperatures. PhIP is metabolized in the liver by cytochrome P450 1A1/2 to carcinogenic metabolite N-hydroxy PhIP, which can form DNA adduct. The ATP binding cassette (ABC) transporters, P-glycoprotein (P-gp), multidrug resistance-associated protein 2 (MRP2) and breast cancer resistance protein (BCRP) are capable of transporting the food-borne procarcinogen PhIP back to the intestinal lumen. In the present study, the uptake and efflux of PhIP were assessed by determining apparent bidirectional permeability coefficients and efflux ratio. The efflux ratio of PhIP with 10 µM caffeic acid was significantly increased compared with control. The mRNA levels of efflux transporters were measured to evaluate the effect of caffeic acid in the presence of PhIP on efflux-mediated transport of PhIP. Caco-2 cells exposed to 10 µM caffeic acid for 3 and 6 h also exhibited higher mRNA levels of P-gp and BCRP than those of control. In contrast, the mRNA level of MRP2 was only slightly induced after 3 h and 6 h. Therefore, caffeic acid at low concentration is expected to be used not only as an antioxidant, but also as an inhibitor of the absorption of food borne carcinogen heterocyclic amines. However, further studies, especially in vivo studies, are required to confirm these results.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
Acido cloridrico, ACS reagent, 37%
Sigma-Aldrich
Cloruro di idrogeno, 4.0 M in dioxane
Sigma-Aldrich
Acido cloridrico, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido cloridrico, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Acido cloridrico, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Cloruro di idrogeno, 2.0 M in diethyl ether
Sigma-Aldrich
Acido cloridrico, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Sigma-Aldrich
Acido cloridrico, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Acido caffeico, ≥98.0% (HPLC)
Sigma-Aldrich
Acido piruvico, 98%
Supelco
Acido cloridrico, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Cloruro di idrogeno, 1.0 M in diethyl ether
Sigma-Aldrich
Acido cloridrico, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%
Sigma-Aldrich
Cloruro di idrogeno, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Acido cloridrico, 32 wt. % in H2O, FCC
Sigma-Aldrich
Acido piruvico, ≥97%, FG
Sigma-Aldrich
Cloruro di idrogeno, 1.0 M in acetic acid
Sigma-Aldrich
Acido piruvico, 95%
Sigma-Aldrich
Acido cloridrico, puriss., 24.5-26.0%
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Acido piruvico, natural, ≥95%, FG
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Acido caffeico, suitable for matrix substance for MALDI-MS, ≥99.0% (HPLC)