Merck
  • Chemical synthesis of the tetrasaccharide repeating unit of the O-polysaccharide isolated from Azospirillum brasilense SR80.
Chemical synthesis of the tetrasaccharide repeating unit of the O-polysaccharide isolated from Azospirillum brasilense SR80.
Carbohydrate research (2015-02-15)
Vikramjit Sarkar, Balaram Mukhopadhyay
ABSTRACT
A linear strategy has been developed for the synthesis of the tetrasaccharide repeating unit of the O-polysaccharide from Azospirillum brasilense SR80. Stepwise glycosylation of the rationally protected thioglycoside donors activated by NIS in the presence of La(OTf)3 furnished the target tetrasaccharide. The glycosylation reactions resulted in the formation of the desired linkage with absolute stereoselectivity and afforded the required derivatives in good to excellent yields. The phthalimido group has been used as the precursor of the desired acetamido group to meet the requirement of 1,2-trans glycosidic linkage.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Cloroformio-d, 99.8 atom % D
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acido acetico, glacial, ReagentPlus®, ≥99%
Sigma-Aldrich
Toluene, ACS reagent, ≥99.5%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Piridina, ACS reagent, ≥99.0%
Sigma-Aldrich
Toluene, suitable for HPLC, 99.9%
Sigma-Aldrich
Cloroformio-d, 99.8 atom % D, contains 0.03 % (v/v) TMS
Sigma-Aldrich
Toluene, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Toluene, anhydrous, 99.8%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Acido acetico, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Piridina, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Piridina, anhydrous, 99.8%
Sigma-Aldrich
Acido acetico, suitable for HPLC
Sigma-Aldrich
Acido acetico, glacial, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8%
Sigma-Aldrich
Acido acetico, glacial, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.8-100.5%
Sigma-Aldrich
Toluene, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.7% (GC)
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
Piridina, ReagentPlus®, ≥99%
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Metanolo, Absolute - Acetone free
Sigma-Aldrich
Cloroformio-d, 99.8 atom % D, contains 0.05 % (v/v) TMS
Sigma-Aldrich
Cloroformio-d, 99.8 atom % D, contains 1 % (v/v) TMS