Merck
  • Effects of an Al(3+)- and Mg(2+)-containing antacid, ferrous sulfate, and calcium carbonate on the absorption of nemonoxacin (TG-873870) in healthy Chinese volunteers.
Effects of an Al(3+)- and Mg(2+)-containing antacid, ferrous sulfate, and calcium carbonate on the absorption of nemonoxacin (TG-873870) in healthy Chinese volunteers.
Acta pharmacologica Sinica (2014-10-21)
Yi-fan Zhang, Xiao-jian Dai, Ting Wang, Xiao-yan Chen, Li Liang, Hua Qiao, Cheng-yuan Tsai, Li-wen Chang, Ping-ting Huang, Chiung-yuan Hsu, Yu-ting Chang, Chen-en Tsai, Da-fang Zhong
ABSTRACT
To evaluate the effects of an Al(3+)- and Mg(2+)-containing antacid, ferrous sulfate, and calcium carbonate on the absorption of nemonoxacin in healthy humans. Two single-dose, open-label, randomized, crossover studies were conducted in 24 healthy male Chinese volunteers (12 per study). In Study 1, the subjects orally received nemonoxacin (500 mg) alone, or an antacid (containing 318 mg of Al(3+) and 496 mg of Mg(2+)) plus nemonoxacin administered 2 h before, concomitantly or 4 h after the antacid. In Study 2, the subjects orally received nemonoxacin (500 mg) alone, or nemonoxacin concomitantly with ferrous sulfate (containing 60 mg of Fe(2+)) or calcium carbonate (containing 600 mg of Ca(2+)). Concomitant administration of nemonoxacin with the antacid significantly decreased the area under the concentration-time curve from time 0 to infinity (AUC0-∞) for nemonoxacin by 80.5%, the maximum concentration (Cmax) by 77.8%, and urine recovery (Ae) by 76.3%. Administration of nemonoxacin 4 h after the antacid decreased the AUC0-∞ for nemonoxacin by 58.0%, Cmax by 52.7%, and Ae by 57.7%. Administration of nemonoxacin 2 h before the antacid did not affect the absorption of nemonoxacin. Administration of nemonoxacin concomitantly with ferrous sulfate markedly decreased AUC0-∞ by 63.7%, Cmax by 57.0%, and Ae by 59.7%, while concomitant administration of nemonoxacin with calcium carbonate mildly decreased AUC0-∞ by 17.8%, Cmax by 14.3%, and Ae by 18.4%. Metal ions, Al(3+), Mg(2+), and Fe(2+) markedly decreased the absorption of nemonoxacin in healthy Chinese males, whereas Ca(2+) had much weaker effects. To avoid the effects of Al(3+) and Mg(2+)-containing drugs, nemonoxacin should be administered ≥2 h before them.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Aluminum oxide, standard grade, Brockmann I, activated, basic
Sigma-Aldrich
Aluminum oxide, activated, Brockmann I, standard grade, neutral
Sigma-Aldrich
Aluminum oxide, powder, primarily α phase, ≤10 μm avg. part. size, 99.5% trace metals basis
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, acidic
Sigma-Aldrich
Aluminum oxide, nanopowder, <50 nm particle size (TEM)
Sigma-Aldrich
Aluminum oxide, powder, 99.99% trace metals basis
Sigma-Aldrich
Aluminum oxide, nanoparticles, <50 nm particle size (DLS), 20 wt. % in isopropanol
Sigma-Aldrich
Aluminum oxide, nanopowder, 13 nm primary particle size (TEM), 99.8% trace metals basis
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, -325 mesh
Sigma-Aldrich
Aluminum oxide, pellets, 3 mm
Sigma-Aldrich
Aluminum oxide, Corundum, α-phase, -100 mesh
Supelco
Aluminum oxide, activated, neutral, Brockmann Activity I
Sigma-Aldrich
Aluminum oxide, fused, powder, primarily α-phase, 100-200 mesh
Sigma-Aldrich
Aluminum oxide, pore size 58 Å, ~150 mesh
Sigma-Aldrich
Aluminum oxide, Type WN-6, Neutral, Activity Grade Super I
Sigma-Aldrich
Aluminum oxide, nanoparticles, 30-60 nm particle size (TEM), 20 wt. % in H2O
Sigma-Aldrich
Aluminum oxide, 99.997% trace metals basis
Sigma-Aldrich
Aluminum oxide, single crystal substrate, <0001>
Sigma-Aldrich
Aluminum oxide, Brockmann I, standard grade, activated, neutral, free-flowing, Redi-Dri
Sigma-Aldrich
Aluminum oxide, mesostructured, MSU-X (wormhole), average pore size 3.8 nm
Supelco
Aluminum oxide, for the determination of hydrocarbons
Sigma-Aldrich
Aluminum oxide, Brockmann I, activated, standard grade, free-flowing, Redi-Dri