Merck
Identification of a naturally occurring retinoid X receptor agonist from Brazilian green propolis.
Biochimica et biophysica acta (2014-06-28)
Ken-Ichi Nakashima, Tohru Murakami, Hiroki Tanabe, Makoto Inoue
ABSTRACT
Brazilian green propolis (BGP), a resinous substance produced from Baccharis dracunculifolia by Africanized honey bees (Apis mellifera), is used as a folk medicine. Our present study explores the retinoid X receptor (RXR) agonistic activity of BGP and the identification of an RXR agonist in its extract. RXRα agonistic activity was evaluated using a luciferase reporter gene assay. Isolation of the RXRα agonist from the ethanolic extract of BGP was performed using successive silica gel and a reversed phase column chromatography. The interaction between the isolated RXRα agonist and RXRα protein was predicted by a receptor-ligand docking simulation. The nuclear receptor (NR) cofactor assay was used to estimate whether the isolated RXRα agonist bound to various NRs, including RXRs and peroxisome proliferator-activated receptors (PPARs). We further examined its effect on adipogenesis in 3T3-L1 fibroblasts. We identified drupanin as an RXRα agonist with an EC50 value of 4.8 ± 1.0 μM. Drupanin activated three RXR subtypes by a similar amount and activated PPARγ moderately. Additionally, drupanin induced adipogenesis and elevated aP2 mRNA levels in 3T3-L1 fibroblasts. Drupanin, a component of BGP, is a novel RXR agonist with slight PPARγ agonistic activity. This study revealed for the first time that BGP activates RXR and drupanin is an RXR agonist in its extract.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Acido retinoico, ≥98% (HPLC), powder
Sigma-Aldrich
Rosiglitazone
Sigma-Aldrich
Bexarotene, ≥98% (HPLC)