Merck
  • A method for long term stabilisation of long chain polyunsaturated fatty acids in dried blood spots and its clinical application.
A method for long term stabilisation of long chain polyunsaturated fatty acids in dried blood spots and its clinical application.
Prostaglandins, leukotrienes, and essential fatty acids (2014-12-03)
Ge Liu, Beverly Sara Mühlhäusler, Robert Alan Gibson
ABSTRACT
Conventional assays of omega-3 long chain polyunsaturated fatty acid (n-3 LCPUFA) status in humans involve venous blood collection and expensive, multi-step processes that limit their usefulness as screening tools. This study aimed to develop a capillary dried blood spot (DBS) system capable of protecting n-3 LCPUFA from oxidation for up to 2 months at room temperature (20-25°C). We demonstrated that a DBS system comprising both an antioxidant and chelating agent on silica-gel coated paper prevented any significant change in the n-3 LCPUFA profile after 2 months. Our DBS assay was then tested in fifty subjects, and this demonstrated the presence of strong and significant correlations between the results obtained from the DBS system and those obtained from conventional measures for all fatty acids, in particular the n-3 LCPUFA EPA and DHA (r>0.96, P<0.0001). This study therefore validates our DBS system as a reliable method for the assessment of n-3 LCPUFA status in humans.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Metanolo, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Acido solforico, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Metanolo, HPLC Plus, ≥99.9%
Sigma-Aldrich
Metanolo, anhydrous, 99.8%
Sigma-Aldrich
Acido citrico, meets analytical specification of Ph. Eur., BP, USP, E330, anhydrous, 99.5-100.5% (based on anhydrous substance)
Sigma-Aldrich
Acido L-ascorbico, powder, suitable for cell culture, γ-irradiated
Sigma-Aldrich
Acido solforico, 99.999%
Sigma-Aldrich
Metanolo, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Metanolo, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Acido citrico, ACS reagent, ≥99.5%
Sigma-Aldrich
Acido L-ascorbico, BioXtra, ≥99.0%, crystalline
Sigma-Aldrich
Metanolo, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., ≥99.8% (GC)
Sigma-Aldrich
2,6-di-tert-butil-4-metilfenolo, ≥99.0% (GC), powder
Sigma-Aldrich
Acido L-ascorbico, suitable for cell culture, suitable for plant cell culture, ≥98%
Sigma-Aldrich
Acido solforico, puriss. p.a., for determination of Hg, ACS reagent, reag. ISO, reag. Ph. Eur., 95.0-97.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Acido L-ascorbico, 99%
Sigma-Aldrich
Acido L-ascorbico, reagent grade, crystalline
Supelco
Acido L-ascorbico, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Metanolo, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Metanolo, ACS reagent, ≥99.8%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Acido citrico, ≥99.5%, FCC, FG
Sigma-Aldrich
Acido citrico, 99%
Supelco
2,6-di-tert-butil-4-metilfenolo, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
USP
Acido ascorbico, United States Pharmacopeia (USP) Reference Standard