Merck
  • Differing coagulation profiles of patients with monoclonal gammopathy of undetermined significance and multiple myeloma.
Differing coagulation profiles of patients with monoclonal gammopathy of undetermined significance and multiple myeloma.
Journal of thrombosis and thrombolysis (2014-10-02)
Maeve P Crowley, Maeve P Crowely, Shane Quinn, Eoin Coleman, Joseph A Eustace, Oonagh M Gilligan, Susan I O'Shea, Susan I O Shea
ABSTRACT
The link between myeloma and thrombosis is well established. Monoclonal gammopathy of undetermined significance (MGUS) has also been associated with an increased risk of thrombosis. It was recently demonstrated that patients with myeloma display changes in thromboelastometry that may indicate a prothrombotic state. There is little data with regard to changes in thromboelastography in patients with myeloma or MGUS. The aim of this study was to investigate the differing coagulation profiles of patients of patients with myeloma and MGUS by means of conventional coagulation tests and thromboelastography. Blood was taken by direct venepuncture from patients with myeloma, MGUS and normal controls. Routine coagulation tests were performed in an accredited hospital laboratory. Thromboelastography (TEG(®)) was performed as per the manufacturer's protocol. Eight patients were recruited in each group. Patients with myeloma had a significantly lower mean haemoglobin level than patients with MGUS or normal controls (p < 0.001). Patients with myeloma had a significantly more prolonged mean prothrombin time than normal controls (p = 0.018) but not patients with MGUS. Patients with myeloma had significantly higher median D-dimer levels than normal controls (p = 0.025), as did patients with MGUS (p = 0.017). Patients with myeloma had a significantly higher mean factor VIII level than normal controls (p = 0.009) and there was a non-significant trend towards patients with MGUS having higher factor VIII levels than normal controls (p = 0.059). There was no significant difference in thromboelastographic parameters between the three groups. Patients with MGUS appear to have a distinct coagulation profile which is intermediate between patients with myeloma and normal controls.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
Cloruro di calcio, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Cloruro di calcio, anhydrous, BioReagent, suitable for insect cell culture, suitable for plant cell culture, ≥96.0%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
Cloruro di calcio, powder, 99.99% trace metals basis
Sigma-Aldrich
Cloruro di calcio
Sigma-Aldrich
Cloruro di calcio, AnhydroBeads, −10 mesh, ≥99.9% trace metals basis
Supelco
Cloruro di calcio, analytical standard, 1.000 g/L Ca2+ in hydrochloric acid, traceable to BAM
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Cloruro di calcio, 0.1 M Ca, analytical standard (for ion-selective electrodes)
Sigma-Aldrich
Cloruro di calcio, AnhydroBeads, −10 mesh, ≥99.99% trace metals basis