Merck
  • Pyrene-based chemosens or detects picric acid upto attogram level through aggregation enhanced excimer emission.
Pyrene-based chemosens or detects picric acid upto attogram level through aggregation enhanced excimer emission.
Analytica chimica acta (2015-03-04)
Rakesh Chopra, Paramjit Kaur, Kamaljit Singh
ABSTRACT
A pyrene-based small molecular weight probe, exhibiting aggregation enhanced excimer emission has been synthesized. The crystalline emissive form detects 2,4,6-trinitrophenol (picric acid) at parts-per-billion concentration in solution and as low as 0.46 attogram in direct contact mode, operating predominantly in a static quenching mechanism, proposed on the basis of steady state and life-time fluorescence measurements.
MATERIALI
Numero di prodotto
Marchio
Descrizione del prodotto
Sigma-Aldrich
1-Aminopyrene, 97%
Prezzi e disponibilità al momento non sono disponibili